Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotinic acid mononucleotide triethylamine | 99.9% | 100 MG
SDP

Catalog No. 5000340993
Encompass_Preferred

Nicotinic acid mononucleotide triethylamine is a key compound formed from nicotinic acid (NA) through the action of nicotinic acid phosphoribosyltransferase during the biosynthesis of NAD+. It also serves as a substrate for nicotinamide mononucleotide/nicotinic acid mononucleotide adenylyltransferase. This product is strictly for research use and is not intended for patient use.

  • Formed from nicotinic acid (NA)
  • Involved in NAD+ biosynthesis pathway
  • Substrate for specific adenylyltransferases
  • White to light yellow solid appearance
  • Molecular weight of 506.97
  • Soluble in DMSO and H2O

Catalog No. 50-003-40993 Supplier Medchemexpress LLC Supplier No. HY128700A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicotinic acid mononucleotide triethylamine is a key compound formed from nicotinic acid (NA) through the action of nicotinic acid phosphoribosyltransferase during the biosynthesis of NAD+. It also serves as a substrate for nicotinamide mononucleotide/nicotinic acid mononucleotide adenylyltransferase. This product is strictly for research use and is not intended for patient use.

  • Formed from nicotinic acid (NA)
  • Involved in NAD+ biosynthesis pathway
  • Substrate for specific adenylyltransferases
  • White to light yellow solid appearance
  • Molecular weight of 506.97
  • Soluble in DMSO and H2O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.