Learn More
Apexbio Technology LLC NICOTINIC-ACID-N-OXIDE-500MG
SDP
Catalog No. 5000569243
Encompass_Preferred
Nicotinic acid N-oxide (CAS No 2398-81-4) is a compound frequently studied in the field of biomedical research particularly in pharmacology and biochemistry Originating from the modification of nicotinic acid it acts through specific biochemical pathways related to nicotinic acid metabolism This compound is known to exhibit activity in the nanomolar to low micromolar range indicating its potential efficacy at relatively low concentrations In vitro it is commonly used in cell-based assays and in vivo studies including animal models to investigate its pharmacokinetic properties and therapeutic potential Additionally it serves as a tool in drug screening to elucidate its effects on various cellular processes Experimental concentrations typically depend on the specific research design taking into account both the target of investigation and the model system employed
Catalog No. 50-005-69243 Supplier Apexbio Technology LLC Supplier No. C7149500
Nicotinic acid N-oxide (CAS No 2398-81-4) is a compound frequently studied in the field of biomedical research particularly in pharmacology and biochemistry Originating from the modification of nicotinic acid it acts through specific biochemical pathways related to nicotinic acid metabolism This compound is known to exhibit activity in the nanomolar to low micromolar range indicating its potential efficacy at relatively low concentrations In vitro it is commonly used in cell-based assays and in vivo studies including animal models to investigate its pharmacokinetic properties and therapeutic potential Additionally it serves as a tool in drug screening to elucidate its effects on various cellular processes Experimental concentrations typically depend on the specific research design taking into account both the target of investigation and the model system employed