Learn More
Medchemexpress LLC Nicotinic acid ribos 5mg
SDP
Catalog No. 5000205211
Encompass_Preferred
Nicotinic acid riboside is a NAD precursor in human cells Nicotinic acid riboside is an authentic intermediate of human NAD metabolism[1][2]
Catalog No. 50-002-05211 Supplier Medchemexpress LLC Supplier No. HYW1043685MG
Nicotinic acid riboside is a NAD precursor in human cells Nicotinic acid riboside is an authentic intermediate of human NAD metabolism[1][2]