Learn More
Medchemexpress LLC HY-W104368 10mg , Nicotinic acid riboside CAS:17720-18-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW10436810MG
Encompass_Preferred
HY-W104368 10mg Nicotinic acid riboside CAS: 17720-18-2 Nicotinic acid riboside is a NAD+ precursor in human cells. Nicotinic acid riboside is an authentic intermediate of human NAD+ metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1603