Learn More
Medchemexpress LLC Nicotinohydrazide | 553-53-7 | 99.35% | 137.14 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW012617100G
Encompass_Preferred
Nicotinohydrazide (Nicotinic acid hydrazide) is a small molecular compound used for research purposes. It exhibits cytotoxic activity and trypanosomicidal activity, making it suitable for various biological studies.
  • Exhibits cytotoxic activity against human HeLa cells (CC50 of 153 μg/mL after 24 hours incubation).
  • Shows trypanosomicidal activity against the amastigote stage of Trypanosoma cruzi Tulahuen C4 in Vero cells (IC50 > 73 μM after 5 days of incubation).
  • Can be stored as powder at -20°C for 3 years or 4°C for 2 years.
  • Can be stored in solvent at -80°C for 2 years or -20°C for 1 year.
  • Soluble in DMSO (100 mg/mL).
Catalog No. 50-004-52928