Learn More
Medchemexpress LLC HY-137394 5mg Medchemexpress, Nicotredole CAS:29876-14-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1373945MG
Encompass_Preferred
Medchemexpress, HY-137394 5mg Nicotredole CAS:29876-14-0 Nicotredole (Tryptamide) is an orally active anti-inflammatory and analgesic agent. Nicotredole exhibits evident antiinflammatory effects of potency comparable with Phenylbutazone. Nicotredole has only weak ulcerogenic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1488