Learn More
Medchemexpress LLC Nicotredole (tryptamide) | 29876-14-0 | MFCD00841846 | 99.9% | C16H15N3O | 10MG
SDP
Catalog No. 5000259087
Encompass_Preferred
Nicotredole (tryptamide) is an orally active small molecule with anti-inflammatory and analgesic activity. It shows anti-inflammatory potency comparable to phenylbutazone while exhibiting only weak ulcerogenic effects, making it useful for preclinical research into inflammation and pain pathways.
  • Orally active anti-inflammatory and analgesic agent.
  • Comparable anti-inflammatory potency to phenylbutazone.
  • Weak ulcerogenic activity relative to reference compounds.
  • High chemical purity (approximately 99.9%).
  • Available as solid and as 10 mM solution in DMSO for in vitro use.
  • Suitable for preclinical pharmacology and mechanism-of-action studies.
Catalog No. 50-002-59087 Supplier Medchemexpress LLC Supplier No. HY13739410MG
Nicotredole (tryptamide) is an orally active small molecule with anti-inflammatory and analgesic activity. It shows anti-inflammatory potency comparable to phenylbutazone while exhibiting only weak ulcerogenic effects, making it useful for preclinical research into inflammation and pain pathways.
  • Orally active anti-inflammatory and analgesic agent.
  • Comparable anti-inflammatory potency to phenylbutazone.
  • Weak ulcerogenic activity relative to reference compounds.
  • High chemical purity (approximately 99.9%).
  • Available as solid and as 10 mM solution in DMSO for in vitro use.
  • Suitable for preclinical pharmacology and mechanism-of-action studies.