Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-nicotyrine | 487-19-4 | MFCD00468104 | 158.20 | C10H10N2 | 10 MG
SDP

Catalog No. 5000226647
Encompass_Preferred

β-Nicotyrine is a minor tobacco alkaloid and a metabolite of nicotine supplied as an analytical standard for research and analytical applications. The compound is suitable for metabolic and analytical studies and is provided in small vial quantities for laboratory use.

  • Used as an analytical standard and research reference material.
  • CAS number 487-19-4.
  • Molecular formula C10H10N2; molecular weight 158.20.
  • Available in small vial pack sizes, for example 10 MG.
  • Recommended storage: solid -20°C (stable for 3 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-26647 Supplier Medchemexpress LLC Supplier No. HY11861410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Nicotyrine is a minor tobacco alkaloid and a metabolite of nicotine supplied as an analytical standard for research and analytical applications. The compound is suitable for metabolic and analytical studies and is provided in small vial quantities for laboratory use.

  • Used as an analytical standard and research reference material.
  • CAS number 487-19-4.
  • Molecular formula C10H10N2; molecular weight 158.20.
  • Available in small vial pack sizes, for example 10 MG.
  • Recommended storage: solid -20°C (stable for 3 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.