Learn More
Medchemexpress LLC Nicur | 2674753-39-8 | 99.1% | C22H16N2O | 10MG
SDP
Catalog No. 5000261754
Encompass_Preferred
NiCur is a potent, selective inhibitor of CBP histone acetyltransferase (HAT) with an IC50 of 0.35 μM. It blocks CBP HAT activity and downregulates p53 activation under genotoxic stress, making it useful for mechanistic cellular and biochemical studies without altering target protein expression.
  • Selective CBP HAT inhibition (IC50 0.35 μM).
  • High purity (99.11%) suitable for research applications.
  • Solid, light yellow to green yellow appearance for easy handling.
  • Soluble at 50 mg/mL in DMSO for in vitro assays.
  • Provides in vivo formulation protocols for dosing studies.
  • Available in multiple small packaging sizes for research use.
Catalog No. 50-002-61754 Supplier Medchemexpress LLC Supplier No. HY13914910MG
NiCur is a potent, selective inhibitor of CBP histone acetyltransferase (HAT) with an IC50 of 0.35 μM. It blocks CBP HAT activity and downregulates p53 activation under genotoxic stress, making it useful for mechanistic cellular and biochemical studies without altering target protein expression.
  • Selective CBP HAT inhibition (IC50 0.35 μM).
  • High purity (99.11%) suitable for research applications.
  • Solid, light yellow to green yellow appearance for easy handling.
  • Soluble at 50 mg/mL in DMSO for in vitro assays.
  • Provides in vivo formulation protocols for dosing studies.
  • Available in multiple small packaging sizes for research use.