Learn More
Medchemexpress LLC Nidogen-1 Mouse P. 100ug
SDP
Catalog No. 5000183462
Encompass_Preferred
Nidogen-1 protein (mouse fragment S428-A665) is a recombinant, N-terminal His-tagged fragment produced in Pichia pastoris and supplied lyophilized. The product is >90% pure by reducing SDS-PAGE, has an apparent molecular mass of ~31 kDa, and endotoxin <1 EU/μg. It is formulated in 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose (pH 8.0); store at -20°C and reconstitute to ≥100 μg/mL before use.
  • Recombinant fragment expressed in Pichia pastoris for eukaryotic glycosylation.
  • N-terminal His tag enables straightforward affinity purification and detection.
  • High purity (>90%) suitable for biochemical and binding assays.
  • Low endotoxin (<1 EU/μg) minimizes immunogenic interference in cell assays.
  • Lyophilized formulation with trehalose for improved stability during storage and shipping.
  • Recommended reconstitution ≥100 μg/mL to maintain protein integrity.
  • Stable long term at -20°C; aliquot after reconstitution for repeated use.
Catalog No. 50-001-83462 Supplier Medchemexpress LLC Supplier No. HYP71763100UG
Nidogen-1 protein (mouse fragment S428-A665) is a recombinant, N-terminal His-tagged fragment produced in Pichia pastoris and supplied lyophilized. The product is >90% pure by reducing SDS-PAGE, has an apparent molecular mass of ~31 kDa, and endotoxin <1 EU/μg. It is formulated in 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose (pH 8.0); store at -20°C and reconstitute to ≥100 μg/mL before use.
  • Recombinant fragment expressed in Pichia pastoris for eukaryotic glycosylation.
  • N-terminal His tag enables straightforward affinity purification and detection.
  • High purity (>90%) suitable for biochemical and binding assays.
  • Low endotoxin (<1 EU/μg) minimizes immunogenic interference in cell assays.
  • Lyophilized formulation with trehalose for improved stability during storage and shipping.
  • Recommended reconstitution ≥100 μg/mL to maintain protein integrity.
  • Stable long term at -20°C; aliquot after reconstitution for repeated use.