Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Nidogen-1/NID1 ELISA Kit

Catalog No. EHNID1X10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHNID1X10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EHNID1X10 Supplier Invitrogen™ Supplier No. EHNID1X10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

The Human Nidogen-1 (Hu NID1) ELISA quantitates Hu NID1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu NID1. Principle of the method The Human NID1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14543
Assay Range 0.123-30 ng/mL
Assay Sensitivity 100 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cell Structure/Organelle Proteins
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1;A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1;; NID
Gene ID (Entrez) 4811
Gene Symbol NID1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases NID-1, Entactin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 2 μL, Serum 2 μL, Supernatant 100 μL
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Nidogen
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.