Learn More
Cayman Chemical Anidulafungin, Purity: ≥95%, CAS: 166663-25-8, 50 mg
SDP
Catalog No. 502983460
Encompass_Preferred
A semisynthetic echinocandin antifungal; inhibits 1,3-β-D glucan synthase from several C. albicans clinical isolates (IC50s = 0.89-7.96 ng/ml for the wild-type enzyme); active against C. albicans, C. parapsilosis, C. tropicalis, and C. guiillermondi (MICs = ≤0.03-4 µg/ml) and A. fumigatus (MIC90 = ≤0.03 µg/ml); increases survival and decreases the severity of pulmonary lesions in A. fumigatus-infected neutropenic rabbits at 10 mg/kg
Catalog No. 50-298-3460 Supplier Cayman Chemical Supplier No. 2075350 MG
A semisynthetic echinocandin antifungal; inhibits 1,3-β-D glucan synthase from several C. albicans clinical isolates (IC50s = 0.89-7.96 ng/ml for the wild-type enzyme); active against C. albicans, C. parapsilosis, C. tropicalis, and C. guiillermondi (MICs = ≤0.03-4 µg/ml) and A. fumigatus (MIC90 = ≤0.03 µg/ml); increases survival and decreases the severity of pulmonary lesions in A. fumigatus-infected neutropenic rabbits at 10 mg/kg