Learn More
Medchemexpress LLC Niemann-Pick C2 (NPC2) protein | >95.0% | 10 UG
SDP
Supplier:  Medchemexpress LLC HYP7469210UG
Encompass_Preferred
Recombinant Niemann-Pick C2 (NPC2) protein (rat) produced in HEK293 cells with a C-terminal His tag. Supplied lyophilized in a 10 μg vial and formulated from PBS (pH 7.4). Designed for research applications investigating intracellular cholesterol trafficking, lipid metabolism, binding interactions, and structural studies.
  • HEK293 expression ensures mammalian glycosylation.
  • C-terminal His tag facilitates purification and detection.
  • High purity (>95%) by reducing SDS-PAGE.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Suitable for biochemical assays, binding studies, and structural work.
  • Available in small quantities for pilot experiments.
Catalog No. 50-002-94258