Learn More
Medchemexpress LLC Niemann-Pick C2 (NPC2) protein | >95.0% | 100 UG
SDP
Supplier:  Medchemexpress LLC HYP74692100UG
Encompass_Preferred
Recombinant rat Niemann-Pick C2 (NPC2) protein (mature E20-G149) expressed in HEK293 cells with a C-terminal 6xHis tag. Supplied as lyophilized powder formulated in PBS (pH 7.4), this protein is intended for research use in biochemical assays, binding studies, and antibody or assay development. It exhibits high purity and low endotoxin suitable for sensitive applications.
Catalog No. 50-002-94322