Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Niemann-Pick C2 (NPC2) protein | >95.0% | 5 UG
SDP

Catalog No. 5000294277
Encompass_Preferred

Recombinant rat Niemann-Pick C2 (NPC2) protein produced in HEK293 cells with a C-terminal 6xHis tag. Supplied lyophilized for biochemical and cell biology research into cholesterol trafficking and lipid metabolism; available in small research quantities such as 5 μg.

  • Recombinant protein expressed in HEK293 cells.
  • C-terminal 6xHis tag for detection and purification.
  • Sequence corresponds to residues E20-G149 of rat NPC2.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Store at -20°C; reconstituted material stable short term at 4°C.

Catalog No. 50-002-94277 Supplier Medchemexpress LLC Supplier No. HYP746925UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat Niemann-Pick C2 (NPC2) protein produced in HEK293 cells with a C-terminal 6xHis tag. Supplied lyophilized for biochemical and cell biology research into cholesterol trafficking and lipid metabolism; available in small research quantities such as 5 μg.

  • Recombinant protein expressed in HEK293 cells.
  • C-terminal 6xHis tag for detection and purification.
  • Sequence corresponds to residues E20-G149 of rat NPC2.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Store at -20°C; reconstituted material stable short term at 4°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.