Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Niemann-Pick C2 (NPC2) protein | >95.0% | 5 UG
SDP

Supplier:  Medchemexpress LLC HYP746945UG

Encompass_Preferred

Recombinant human Niemann-Pick C2 (NPC2) protein expressed in HEK293 cells, supplied as a C-terminal His-tagged, lyophilized powder for use in research on cholesterol trafficking and lipid metabolism.

  • Purity >95% by Tris-Bis PAGE.
  • Expression system: HEK293 mammalian cells.
  • Contains C-terminal His tag for detection or purification.
  • Supplied as 5 μg lyophilized powder.
  • Endotoxin <1 EU/μg (LAL assay).
  • Glycosylated mature protein, full-length isoform-1.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Store at -20°C; shelf life 2 years (unreconstituted); aliquot for long-term storage.

Catalog No. 50-002-94286


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.