Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Niemann-Pick C2 (NPC2) protein, human (HEK293, C-His) | >95.0% | 50 UG
SDP

Catalog No. 5000294307
Encompass_Preferred

Recombinant human Niemann-Pick C2 (NPC2) protein expressed in HEK293 cells, supplied as a lyophilized powder with a C-terminal His tag. Formulated from a 0.22 μm filtered PBS solution (pH 7.4) and provided in a 50 μg vial, it is intended for biochemical and biophysical studies of cholesterol trafficking and protein-lipid interactions. For research use only.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4.
  • Purity >95.0% as determined by Tris-Bis PAGE.
  • Supplied as a 50 μg vial for lab-scale experiments.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • Suitable for biochemical assays, SPR, and protein interaction studies.

Catalog No. 50-002-94307 Supplier Medchemexpress LLC Supplier No. HYP7469450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Niemann-Pick C2 (NPC2) protein expressed in HEK293 cells, supplied as a lyophilized powder with a C-terminal His tag. Formulated from a 0.22 μm filtered PBS solution (pH 7.4) and provided in a 50 μg vial, it is intended for biochemical and biophysical studies of cholesterol trafficking and protein-lipid interactions. For research use only.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4.
  • Purity >95.0% as determined by Tris-Bis PAGE.
  • Supplied as a 50 μg vial for lab-scale experiments.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • Suitable for biochemical assays, SPR, and protein interaction studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.