Learn More
Medchemexpress LLC Niemann-Pick C2 (NPC2) protein, human recombinant | >95.0% | 10 UG
SDP
Catalog No. 5000294538
Encompass_Preferred
Recombinant human Niemann-Pick C2 (NPC2) protein expressed in HEK293 cells and supplied with a C-terminal His tag. The protein corresponds to the mature sequence (E20-L151) and is supplied as a purified reagent for research applications involving cholesterol trafficking and lysosomal protein function.
Catalog No. 50-002-94538 Supplier Medchemexpress LLC Supplier No. HYP7469410UG
Recombinant human Niemann-Pick C2 (NPC2) protein expressed in HEK293 cells and supplied with a C-terminal His tag. The protein corresponds to the mature sequence (E20-L151) and is supplied as a purified reagent for research applications involving cholesterol trafficking and lysosomal protein function.