Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Niemann-Pick C2 (NPC2) protein | >95.0% | 10 UG
SDP

Catalog No. 5000294244
Encompass_Preferred

Recombinant mouse Niemann-Pick C2 (NPC2) protein expressed in HEK293 cells and supplied as a lyophilized powder. The protein carries a C-terminal His tag, has a molecular weight of approximately 15.9-19 kDa, and is >95.0% pure by reducing SDS-PAGE. Formulated from 0.22 μm filtered PBS (pH 7.4) and recommended for biochemical assays, binding studies, and assay development.

  • Recombinant protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • High purity (>95.0%) for reliable experimental results.
  • Lyophilized formulation for improved shelf life and transport.
  • Buffered in PBS, pH 7.4, for straightforward reconstitution.
  • Recommended storage at -20°C; aliquot and freeze for extended storage.
  • Suitable for biochemical characterization and binding studies.

Catalog No. 50-002-94244 Supplier Medchemexpress LLC Supplier No. HYP7469310UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Niemann-Pick C2 (NPC2) protein expressed in HEK293 cells and supplied as a lyophilized powder. The protein carries a C-terminal His tag, has a molecular weight of approximately 15.9-19 kDa, and is >95.0% pure by reducing SDS-PAGE. Formulated from 0.22 μm filtered PBS (pH 7.4) and recommended for biochemical assays, binding studies, and assay development.

  • Recombinant protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • High purity (>95.0%) for reliable experimental results.
  • Lyophilized formulation for improved shelf life and transport.
  • Buffered in PBS, pH 7.4, for straightforward reconstitution.
  • Recommended storage at -20°C; aliquot and freeze for extended storage.
  • Suitable for biochemical characterization and binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.