Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Niemann Pick C2 (NPC2) protein, recombinant human | >95.0% | 100 UG
SDP

Catalog No. 5000294480
Encompass_Preferred

Recombinant human Niemann Pick C2 (NPC2) protein produced in HEK293 cells and C-terminally His-tagged. Supplied lyophilized, this protein is intended for in vitro research into cholesterol trafficking and lipid metabolism, and is provided with sequence and purity information for experimental planning.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag facilitates purification and detection.
  • Full-length mature sequence (E20-L151) provided.
  • Purity greater than 95% as determined by Tris-Bis PAGE.
  • Lyophilized from PBS, pH 7.4.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O.
  • Stable for long-term storage at -20°C to -80°C; add carrier protein for best recovery.
  • Suitable for biochemical assays and functional studies of lipid transport.

Catalog No. 50-002-94480 Supplier Medchemexpress LLC Supplier No. HYP74694100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Niemann Pick C2 (NPC2) protein produced in HEK293 cells and C-terminally His-tagged. Supplied lyophilized, this protein is intended for in vitro research into cholesterol trafficking and lipid metabolism, and is provided with sequence and purity information for experimental planning.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag facilitates purification and detection.
  • Full-length mature sequence (E20-L151) provided.
  • Purity greater than 95% as determined by Tris-Bis PAGE.
  • Lyophilized from PBS, pH 7.4.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O.
  • Stable for long-term storage at -20°C to -80°C; add carrier protein for best recovery.
  • Suitable for biochemical assays and functional studies of lipid transport.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.