Learn More
Medchemexpress LLC Niemann-Pick C2 protein (human) | >95.0% | 50 UG
SDP
Catalog No. 5000294535
Encompass_Preferred
Recombinant human Niemann-Pick C2 (NPC2) protein expressed in HEK293 cells with a His-tag, supplied as purified recombinant protein for research use. NPC2 is a lysosomal cholesterol-binding protein involved in intracellular cholesterol trafficking and lipid metabolism, and is suitable for biochemical assays, structural studies, and antibody generation.
  • Human sequence and mammalian expression for native glycosylation.
  • His tag for affinity purification and detection.
  • Purified, >95% as determined by reducing SDS-PAGE.
  • Supplied in a buffer compatible with common downstream assays and storage.
  • Intended for research use only; not for therapeutic or diagnostic use.
Catalog No. 50-002-94535 Supplier Medchemexpress LLC Supplier No. HYP7469250UG
Recombinant human Niemann-Pick C2 (NPC2) protein expressed in HEK293 cells with a His-tag, supplied as purified recombinant protein for research use. NPC2 is a lysosomal cholesterol-binding protein involved in intracellular cholesterol trafficking and lipid metabolism, and is suitable for biochemical assays, structural studies, and antibody generation.
  • Human sequence and mammalian expression for native glycosylation.
  • His tag for affinity purification and detection.
  • Purified, >95% as determined by reducing SDS-PAGE.
  • Supplied in a buffer compatible with common downstream assays and storage.
  • Intended for research use only; not for therapeutic or diagnostic use.