Learn More
Selleck Chemical LLC Nifedipine BAY-a-1040-50mg
SDP
Supplier:  Selleck Chemical LLC S180850MG
Encompass_Preferred
Nifedipine (BAY-a-1040) is a dihydropyridine calcium channel blocker, used to lower hypertension and to treat angina.
Catalog No. 50-238-9592