Learn More
Medchemexpress LLC Nifedipine-d6 (BAY-a-1040-d6) | 1188266-14-9 | 1 MG
SDP
Catalog No. 5000405834
Encompass_Preferred
Nifedipine-d6 (BAY-a-1040-d6) is a deuterium labeled nifedipine, which acts as a potent calcium channel blocker. It is intended for research use only. Deuteration, the incorporation of stable heavy isotopes of hydrogen into drug molecules, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05834 Supplier Medchemexpress LLC Supplier No. HYB0284S1MG
Nifedipine-d6 (BAY-a-1040-d6) is a deuterium labeled nifedipine, which acts as a potent calcium channel blocker. It is intended for research use only. Deuteration, the incorporation of stable heavy isotopes of hydrogen into drug molecules, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS