Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Nifedipine USP
SDP

Catalog No. 14185428 Shop All Medisca Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-428 25 g
14-185-622 500 g
14-185-283 5 g
14-185-285 100 g
14-185-402 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-185-428 Supplier Medisca Supplier No. 028004
Get 1 Extra Chemical When You Buy 4
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nifedipine USP is a yellow powder. It is affected by exposure to light.

Nifedipine USP is a yellow powder. It is affected by exposure to light.

Specifications

Chemical Name or Material Nifedipine USP
Packaging Jar, 50mL, White, HDPE, Round, 38-400, TE Pano Neck
CAS 21829-25-4
Type Drug Standard
Molecular Weight (g/mol) 346.3372
Quantity 25 g
Percent Purity 98 to 102 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.