Learn More
Cayman Chemical Nifekalant (hydrochloride), Purity: ≥98%, CAS: 130656-51-8, 10 mg
SDP
Catalog No. 502983862
Encompass_Preferred
A class III antiarrhythmic agent; inhibits IKr (IC50 = 10 µM); prolongs cardiac APD in isolated dog Purkinje fibers at 1 and 3 µg/ml; inhibits PES-induced arrhythmias in dogs post myocardial infarction; increases atrial and ventricular ERPs in anesthetized dogs; inhibits MATE1 (IC50 = 6.5 µM)
Catalog No. 50-298-3862 Supplier Cayman Chemical Supplier No. 2099710 MG
A class III antiarrhythmic agent; inhibits IKr (IC50 = 10 µM); prolongs cardiac APD in isolated dog Purkinje fibers at 1 and 3 µg/ml; inhibits PES-induced arrhythmias in dogs post myocardial infarction; increases atrial and ventricular ERPs in anesthetized dogs; inhibits MATE1 (IC50 = 6.5 µM)