Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nifenalol hydrochloride | 5704-60-9 | 260.72 | C11H17ClN2O3 | 5 MG
SDP

Catalog No. 5000263218
Encompass_Preferred

Nifenalol hydrochloride is a β-adrenergic receptor antagonist used in cardiac electrophysiology and pharmacology research. The compound is reported to induce early afterdepolarization (EAD) and is used to study arrhythmia and blood pressure-related effects.

  • Beta-adrenergic receptor antagonist activity
  • Induces early afterdepolarization useful for arrhythmia studies
  • Hydrochloride salt form for improved solubility
  • Molecular formula C11H17ClN2O3
  • Molecular weight 260.72
  • Available in small research pack sizes (e.g., 5 mg)
  • Intended for in vitro and in vivo research applications

Catalog No. 50-002-63218 Supplier Medchemexpress LLC Supplier No. HY1009525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nifenalol hydrochloride is a β-adrenergic receptor antagonist used in cardiac electrophysiology and pharmacology research. The compound is reported to induce early afterdepolarization (EAD) and is used to study arrhythmia and blood pressure-related effects.

  • Beta-adrenergic receptor antagonist activity
  • Induces early afterdepolarization useful for arrhythmia studies
  • Hydrochloride salt form for improved solubility
  • Molecular formula C11H17ClN2O3
  • Molecular weight 260.72
  • Available in small research pack sizes (e.g., 5 mg)
  • Intended for in vitro and in vivo research applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.