Learn More
Medchemexpress LLC Nifenalol hydrochloride | 5704-60-9 | 99.9% | 260.72 | C11H17ClN2O3 | 25 MG
SDP
Catalog No. 5000212245
Encompass_Preferred
Nifenalol hydrochloride is a β-adrenergic receptor antagonist supplied as a white to off-white solid for research use. It is used in pharmacology and cardiac electrophysiology studies, including investigation of early afterdepolarization (EAD) effects. The compound is provided in milligram-scale packs and has manufacturer guidance for solid and solution storage.
  • β-adrenergic receptor antagonist for pharmacological research.
  • Useful for studying cardiac electrophysiology and EAD mechanisms.
  • High reported purity suitable for laboratory assays (99.9%).
  • White to off-white solid, stable under recommended storage.
  • Available in milligram-scale pack sizes for small-scale experiments.
Catalog No. 50-002-12245 Supplier Medchemexpress LLC Supplier No. HY10095225MG
Nifenalol hydrochloride is a β-adrenergic receptor antagonist supplied as a white to off-white solid for research use. It is used in pharmacology and cardiac electrophysiology studies, including investigation of early afterdepolarization (EAD) effects. The compound is provided in milligram-scale packs and has manufacturer guidance for solid and solution storage.
  • β-adrenergic receptor antagonist for pharmacological research.
  • Useful for studying cardiac electrophysiology and EAD mechanisms.
  • High reported purity suitable for laboratory assays (99.9%).
  • White to off-white solid, stable under recommended storage.
  • Available in milligram-scale pack sizes for small-scale experiments.