Learn More
Medchemexpress LLC Nifenalol hydrochloride | 5704-60-9 | 99.9% | 260.72 g/mol | C11H17ClN2O3 | 1 ML
SDP
Catalog No. 5000212244
Encompass_Preferred
Nifenalol hydrochloride is a β-adrenergic receptor antagonist used in cardiovascular and cardiac electrophysiology research. The product is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is accompanied by standard documentation for handling and quality verification.
  • β-adrenergic receptor antagonist for cardiac research.
  • High purity suitable for research applications.
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • White to off-white solid appearance when provided as a solid.
  • Includes data sheet, COA, and SDS for safety and compliance.
  • Storage recommendations provided for solid and solution forms.
Catalog No. 50-002-12244 Supplier Medchemexpress LLC Supplier No. HY10095210MM 1ML
Nifenalol hydrochloride is a β-adrenergic receptor antagonist used in cardiovascular and cardiac electrophysiology research. The product is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is accompanied by standard documentation for handling and quality verification.
  • β-adrenergic receptor antagonist for cardiac research.
  • High purity suitable for research applications.
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • White to off-white solid appearance when provided as a solid.
  • Includes data sheet, COA, and SDS for safety and compliance.
  • Storage recommendations provided for solid and solution forms.