Learn More
Medchemexpress LLC Nifenalol hydrochloride | 5704-60-9 | 99.9% | 260.72 g/mol | C11H17ClN2O3 | 10 MG
SDP
Catalog No. 5000212634
Encompass_Preferred
Nifenalol hydrochloride is a β-adrenergic receptor antagonist used in pharmacological research. It is supplied as a solid and as a pre-made DMSO solution for convenient dosing and assay preparation, and is commonly used in biochemical and electrophysiological studies of β-adrenoceptor activity.
  • High purity (99.9%) suitable for biochemical and cellular assays.
  • Available in small solid quantities and as 10 mM solutions in DMSO for convenience.
  • Supports studies of β-adrenergic receptor antagonism and early afterdepolarization effects.
  • Characterized by CAS number 5704-60-9 and standard chemical identifiers.
  • Supplied for research use with accompanying datasheet information for handling.
Catalog No. 50-002-12634 Supplier Medchemexpress LLC Supplier No. HY10095210MG
Nifenalol hydrochloride is a β-adrenergic receptor antagonist used in pharmacological research. It is supplied as a solid and as a pre-made DMSO solution for convenient dosing and assay preparation, and is commonly used in biochemical and electrophysiological studies of β-adrenoceptor activity.
  • High purity (99.9%) suitable for biochemical and cellular assays.
  • Available in small solid quantities and as 10 mM solutions in DMSO for convenience.
  • Supports studies of β-adrenergic receptor antagonism and early afterdepolarization effects.
  • Characterized by CAS number 5704-60-9 and standard chemical identifiers.
  • Supplied for research use with accompanying datasheet information for handling.