Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nifenalol hydrochloride | 5704-60-9 | 99.9% | 260.72 g/mol | C11H17ClN2O3 | 100 MG
SDP

Catalog No. 5000212628
Encompass_Preferred

Nifenalol hydrochloride is a research-grade β-adrenergic receptor antagonist (hydrochloride salt) used in pharmacology and cardiac electrophysiology studies. It is provided as a high-purity solid for biochemical and cellular assays, with recommended storage and solvent stability guidance supplied by the manufacturer.

  • Chemical identity: CAS number 5704-60-9; formula C11H17ClN2O3; molecular weight 260.72 g/mol.
  • Primary activity: β-adrenergic receptor antagonist demonstrating early afterdepolarization effects.
  • High purity: 99.9% as listed by manufacturer.
  • Available formats: solid and solution stocks, sold in multiple sizes including 100 MG.
  • Storage recommendation: sealed, away from moisture and light; solids refrigerated at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-12628 Supplier Medchemexpress LLC Supplier No. HY100952100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nifenalol hydrochloride is a research-grade β-adrenergic receptor antagonist (hydrochloride salt) used in pharmacology and cardiac electrophysiology studies. It is provided as a high-purity solid for biochemical and cellular assays, with recommended storage and solvent stability guidance supplied by the manufacturer.

  • Chemical identity: CAS number 5704-60-9; formula C11H17ClN2O3; molecular weight 260.72 g/mol.
  • Primary activity: β-adrenergic receptor antagonist demonstrating early afterdepolarization effects.
  • High purity: 99.9% as listed by manufacturer.
  • Available formats: solid and solution stocks, sold in multiple sizes including 100 MG.
  • Storage recommendation: sealed, away from moisture and light; solids refrigerated at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.