Learn More
Cayman Chemical NIfenalolhydrochlorIde 10mg
SDP
Catalog No. 502946588
Encompass_Preferred
A β-AR antagonist; inhibits isoproterenol-induced contractions in isolated rabbit right ventricular strips (pA2 = 6.05)
Catalog No. 50-294-6588 Supplier Cayman Chemical Supplier No. 3294410 MG
A β-AR antagonist; inhibits isoproterenol-induced contractions in isolated rabbit right ventricular strips (pA2 = 6.05)