Learn More
Cayman Chemical NIfuroxazIde-d4 500ug
SDP
Catalog No. 502948278
Encompass_Preferred
An internal standard for the quantification of nifuroxazide by GC- or LC-MS
Catalog No. 50-294-8278 Supplier Cayman Chemical Supplier No. 33382500 UG
An internal standard for the quantification of nifuroxazide by GC- or LC-MS