Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nifuroxime, 99%, Thermo Scientific™

Catalog No. AC165141000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC165141000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC165141000 Supplier Thermo Scientific Chemicals Supplier No. 165141000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 555-15-7
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C5H4N2O4
MDL Number MFCD00198004,MFCD00198004
Quantity 100g
Merck Index 10, 6384
Synonym nifuroxime, 5-nitro-2-furaldehyde oxime, usaf ea-5, 2-furaldehyde, 5-nitro-, oxime, 2-furaldoxime, 5-nitro, 5-nitro-2-furaldoxime, carbazylquinone, carbilquinone, 5-nitrofuran-2-carbaldehyde oxime, 5-nitro-2-furancarboxaldehyde oxime
InChI Key PTBKFATYSVLSSD-ZZXKWVIFSA-N
SMILES O\N=C\C1=CC=C(O1)[N+]([O-])=O
IUPAC Name (E)-N-[(5-nitrofuran-2-yl)methylidene]hydroxylamine
Molecular Weight (g/mol) 156.10
PubChem CID 6478035
Formula Weight 156.1
Percent Purity ≥98.5%
Chemical Name or Material Nifuroxime, 99%
Show More Show Less
Hazard Category LIGHT SENSITIVE
EINECSNumber 209-082-4
RTECSNumber LT7525000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.