Learn More
Medchemexpress LLC Nifurpirinol | 13411-16-0 | 98.7% | C12H10N2O4 | 10MG
SDP
Catalog No. 5000260784
Encompass_Preferred
Nifurpirinol is a nitroaromatic compound used as a pro-substrate for bacterial nitroreductase (NTR) in research applications. NTR enzymatic reduction of nifurpirinol produces cytotoxic metabolites that enable selective ablation of NTR-expressing cells in model organisms and cell systems. This reagent is supplied at high purity for analytical and experimental use and is intended for research use only.
  • Selective NTR substrate for targeted cell ablation.
  • Enables regeneration and disease modeling in vivo.
  • Applicable to studies of pancreatic β cell, osteoblast, dopaminergic neuron, and podocyte ablation.
  • High purity suitable for analytical and research applications.
  • Provided with authoritative chemical identifiers for traceability.
Catalog No. 50-002-60784 Supplier Medchemexpress LLC Supplier No. HY13547010MG
Nifurpirinol is a nitroaromatic compound used as a pro-substrate for bacterial nitroreductase (NTR) in research applications. NTR enzymatic reduction of nifurpirinol produces cytotoxic metabolites that enable selective ablation of NTR-expressing cells in model organisms and cell systems. This reagent is supplied at high purity for analytical and experimental use and is intended for research use only.
  • Selective NTR substrate for targeted cell ablation.
  • Enables regeneration and disease modeling in vivo.
  • Applicable to studies of pancreatic β cell, osteoblast, dopaminergic neuron, and podocyte ablation.
  • High purity suitable for analytical and research applications.
  • Provided with authoritative chemical identifiers for traceability.