Learn More
Medchemexpress LLC Nifursol-15N2,d2 | 1246833-64-6 | 369.21 g/mol | C12H5D2N3^15N2O9 | 1mg
SDP
Catalog No. 5000270138
Encompass_Preferred
Nifursol-15N2 d2 is the deuterium and 15N-labeled Nifursol (HY-B1703) Nifursol is an orally active veterinary antibiotic Nifursol inhibits Histomonas meleagridis Nifursol prevents histomoniasis in turkeys Nifursol is widely used in studies of Escherichia coli gastroenteritis in Salmonella-infected poultry and aquatic animals[1][2][3]
Catalog No. 50-002-70138 Supplier Medchemexpress LLC Supplier No. HYW7779221MG
Nifursol-15N2 d2 is the deuterium and 15N-labeled Nifursol (HY-B1703) Nifursol is an orally active veterinary antibiotic Nifursol inhibits Histomonas meleagridis Nifursol prevents histomoniasis in turkeys Nifursol is widely used in studies of Escherichia coli gastroenteritis in Salmonella-infected poultry and aquatic animals[1][2][3]