Learn More
TARGETMOL CHEMICALS INC NIFURTIMOX 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T532510MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nifurtimox (BAY-A-2502) is an antiprotozoal agent (IC50s = 9.91 12.28 and 10.44 uM against Taluahuén LQ and Brener strains of T. cruzi- epimastigotes respectively). purity: 99%
Catalog No. 50-257-9789