Learn More
Medchemexpress LLC HY-W040073 10mg Medchemexpress, Nifurtimox CAS:23256-30-6 Purity:>98%
SDP
Catalog No. 501934654
Encompass_Preferred
Medchemexpress, HY-W040073 10mg Nifurtimox CAS:23256-30-6 Nifurtimox, an antiprotozoal agent, which is generally used for the treatment of infections with Trypanosoma cruzi, has been used in the therapy of neuroblastoma. Nifurtimox affects enzyme activity of lactate dehydrogenase ( LDH ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4654 Supplier Medchemexpress LLC Supplier No. HYW04007310MG
Medchemexpress, HY-W040073 10mg Nifurtimox CAS:23256-30-6 Nifurtimox, an antiprotozoal agent, which is generally used for the treatment of infections with Trypanosoma cruzi, has been used in the therapy of neuroblastoma. Nifurtimox affects enzyme activity of lactate dehydrogenase ( LDH ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.