Learn More
Cayman Chemical Nifurtimox-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502937486
Encompass_Preferred
An internal standard for the quantification of nifurtimox by GC- or LC-MS
Catalog No. 50-293-7486 Supplier Cayman Chemical Supplier No. 288891 MG
An internal standard for the quantification of nifurtimox by GC- or LC-MS