Learn More
TARGETMOL CHEMICALS INC Nigericin sodium salt, 100mg | Cas No. 28643-80-3 | Purity: 99.52%
SDP
Catalog No. NC3918287
Encompass_Preferred
Nigericin sodium salt is an antibiotic, an NLRP3 activator, and a cationic ion carrier. Nigericin sodium triggers the activation of the NALP3 inflammasome, which inhibits Golgi function and suppresses the growth of Gram-positive bacteria.
Catalog No. NC3918287 Supplier TARGETMOL CHEMICALS INC Supplier No. T3092100MG
Nigericin sodium salt is an antibiotic, an NLRP3 activator, and a cationic ion carrier. Nigericin sodium triggers the activation of the NALP3 inflammasome, which inhibits Golgi function and suppresses the growth of Gram-positive bacteria.