Learn More
Apexbio Technology LLC Nigericin sodium salt, 10mg CAS# 28643-80-3
SDP
Supplier:  Apexbio Technology LLC B764410
Encompass_Preferred
Nigericin sodium salt is an ionophore that exchanges K+ for H+ across biological membranes.Ionophore is a lipid-soluble molecule that transports ions across the cell membrane.In medium with Na+, nigericin scarcely affected ADP-induced platelets aggregation, and slightly inhibited thrombin-induced platelets aggregation. Also, nigericin decreased the cytoplasmic pH in the medium with Na+. The effects of nigericin on platelet aggregation are mainly due to its effects on the cytoplasmic pH [1]. The K+ ionophore nigericin is highly effective as an ionophore for Pb2+. Physiological concentrations of Ca2+ or Mg2+ didnt inhibit nigericin-catalyzed Pb2+ transport, while K+ and Na+ concentrations (0-100 mM) modestly affected Pb2+ transport. Nigericin may be helpful in the treatment of Pb intoxication. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8380