Learn More
Medchemexpress LLC HY-100381 10mg , Nigericin (sodium salt) CAS:28643-80-3 Purity:>98%
SDP
Catalog No. 502260362
Encompass_Preferred
HY-100381 10mg Nigericin (sodium salt) CAS: 28643-80-3 Nigericin sodium salt is an antibiotic from Streptomyces hygroscopicus that works by acting as an H+, K+, and Pb2+ ionophore, a NLRP3 activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0362 Supplier Medchemexpress LLC Supplier No. HY10038110MG
HY-100381 10mg Nigericin (sodium salt) CAS: 28643-80-3 Nigericin sodium salt is an antibiotic from Streptomyces hygroscopicus that works by acting as an H+, K+, and Pb2+ ionophore, a NLRP3 activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.