Learn More
TARGETMOL CHEMICALS INC NIGERICIN SODIUM SALT 10MG
SDP
Catalog No. 502577004
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Nigericin sodium salt is an antibiotic from Streptomyces hygroscopicus and is a NLRP3 activator.Nigericin sodium is a cationic ionophore that inhibits Golgi function and suppresses growth of gram positive bacteria. It also prevents viral activation and triggers activation of the NALP3 inflammasome. purity: 99%
Catalog No. 50-257-7004 Supplier TARGETMOL CHEMICALS INC Supplier No. T309210MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Nigericin sodium salt is an antibiotic from Streptomyces hygroscopicus and is a NLRP3 activator.Nigericin sodium is a cationic ionophore that inhibits Golgi function and suppresses growth of gram positive bacteria. It also prevents viral activation and triggers activation of the NALP3 inflammasome. purity: 99%