Learn More
Ubpbio An ionophore that disrupts mitochondrial membrane potential. Often used for activation of the NLRP3 inflammasome. CAS# 28643-80-3, > 95% purity
Catalog No. 502339362
Encompass_Preferred
Nigericin sodium salt is an antibiotic derived from Streptomyces hygroscopicus. Functions as an H+, K+, and Pb2+ ionophore. Disrupts mitochondrial membrane potential and activates the NLRP3 inflammasome to induce inflammation.
Catalog No. 50-233-9362 Supplier Ubpbio Supplier No. F320025
Nigericin sodium salt is an antibiotic derived from Streptomyces hygroscopicus. Functions as an H+, K+, and Pb2+ ionophore. Disrupts mitochondrial membrane potential and activates the NLRP3 inflammasome to induce inflammation.