Learn More
Medchemexpress LLC Nigericin sodium salt | 28643-80-3 | 746.94 | C40H67NaO11 | 100 MG
SDP
Catalog No. 5000212737
Encompass_Preferred
Nigericin sodium salt is a polyether ionophore antibiotic used as a research reagent and NLRP3 inflammasome activator. Supplied as an analytical standard for laboratory and biochemical research, it is commonly used to manipulate intracellular ion gradients, study inflammasome activation, and probe mitochondrial ion transport. Not for human use.
  • Acts as a K+/H+ ionophore to alter intracellular ion gradients.
  • Used to activate the NLRP3 inflammasome in cell-based assays.
  • Provided as an analytical standard for research applications.
  • Available in multiple pack sizes, including 100 mg.
  • Stable under recommended sealed storage; follow the certificate of analysis for conditions.
Catalog No. 50-002-12737 Supplier Medchemexpress LLC Supplier No. HY100381100MG
Nigericin sodium salt is a polyether ionophore antibiotic used as a research reagent and NLRP3 inflammasome activator. Supplied as an analytical standard for laboratory and biochemical research, it is commonly used to manipulate intracellular ion gradients, study inflammasome activation, and probe mitochondrial ion transport. Not for human use.
  • Acts as a K+/H+ ionophore to alter intracellular ion gradients.
  • Used to activate the NLRP3 inflammasome in cell-based assays.
  • Provided as an analytical standard for research applications.
  • Available in multiple pack sizes, including 100 mg.
  • Stable under recommended sealed storage; follow the certificate of analysis for conditions.