Learn More
Selleck Chemical LLC Nigericin sodium salt
SDP
Catalog No. 5000002702
Encompass_Preferred
Nigericin sodium salt is an antibiotic derived from Streptomyces hygroscopicus that works by acting as an H K and Pb2 ionophore Nigericin can activate NLRP3 inflammasome to induce pro-inflammatory and immunostimulatory processes
Catalog No. 50-000-02702 Supplier Selleck Chemical LLC Supplier No. S66531G
Nigericin sodium salt is an antibiotic derived from Streptomyces hygroscopicus that works by acting as an H K and Pb2 ionophore Nigericin can activate NLRP3 inflammasome to induce pro-inflammatory and immunostimulatory processes