Learn More
AdipoGen Nigericin . sodium salt,Chemical. CAS: 28643-80-3.
SDP
Catalog No. 50464327
Encompass_Preferred
Nigericin . sodium salt, AdipoGen Life Sciences. Chemical. CAS: 28643-80-3. Formula: C40H67O11 . Na. MW: 724.0 . 23.0. Isolated from Streptomyces hygroscopicus. Antibiotic. High affinity ionophore for monovalent cations such as H+, K+, Na+, Pb2+. Shows antibacterial (Gram-positive), antifungal, antitumor and antiviral activity. Disrupts membrane potential of mitochondria. NLRP3/NALP3 activator. Signals through pannexin-1 to induce caspase-1 maturation and IL-1beta processing and release. Autophagy modulator
Catalog No. 50-464-327 Supplier AdipoGen Supplier No. AGCN20020M005
Nigericin . sodium salt, AdipoGen Life Sciences. Chemical. CAS: 28643-80-3. Formula: C40H67O11 . Na. MW: 724.0 . 23.0. Isolated from Streptomyces hygroscopicus. Antibiotic. High affinity ionophore for monovalent cations such as H+, K+, Na+, Pb2+. Shows antibacterial (Gram-positive), antifungal, antitumor and antiviral activity. Disrupts membrane potential of mitochondria. NLRP3/NALP3 activator. Signals through pannexin-1 to induce caspase-1 maturation and IL-1beta processing and release. Autophagy modulator