Learn More
Enzo Life Sciences Nigericin. sodium salt (5mg). CAS: 28643-80-3; 28380-24-7
SDP
Catalog No. 502010397
Encompass_Preferred
Acts as an H+, K+, Na+, Pb2+ ionophore. Is most commonly an antiporter of H+ and K+. Stimulates Ca2+ release from mitochondrial stores by disruption of membrane potantial. Inhibits the Golgi functions. Activator of NALP3. Alternative name: Antibiotic K 178. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml, heating may be required), dichloromethane (10mg/ml), methanol (10mg/ml), ethanol (5mg/ml) and ethyl acetate. Long Term Storage: -20°C.
Catalog No. 50-201-0397 Supplier Enzo Life Sciences Supplier No. BMLCA4210005
Acts as an H+, K+, Na+, Pb2+ ionophore. Is most commonly an antiporter of H+ and K+. Stimulates Ca2+ release from mitochondrial stores by disruption of membrane potantial. Inhibits the Golgi functions. Activator of NALP3. Alternative name: Antibiotic K 178. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml, heating may be required), dichloromethane (10mg/ml), methanol (10mg/ml), ethanol (5mg/ml) and ethyl acetate. Long Term Storage: -20°C.