Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nigericin Sodium Salt, G-Biosciences
SCI_ED

Catalog No. S139765
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139765 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139765 Supplier G-Biosciences Supplier No. RC1065
Add to Cart
Add to Cart

Nigericin is an ionophore antibiotic isolated from Streptomyces hygroscopicus. It has been shown to suppress DNA synthesis in Ehrlich ascites carcinoma.

Nigericin is insoluble in water, but soluble in methanol and chloroform. Nigericin sodium disrupts membrane potential by acting as an antiporter of H+ and K+ ions by exchanging K+ for H+. This causes a rapid decrease in intracellular pH.

Chemical Identifiers

Molecular Weight (g/mol) 746.94 g/mol

Specifications

Boiling Point 779.9°C
Quantity 50 mg
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.