Learn More
Electron Microscopy Sciences NIGHTSEA™ Barrier Filter Glasses, Cyan
SDP
Catalog No. 501927549
Encompass_Preferred
Glasses for use with the NIGHTSEA BlueStar flashlight meet ANSI Z87.1 impact standards for safety glasses.
Color: Cyan
Catalog No. 50-192-7549 Supplier Electron Microscopy Sciences Supplier No. FGCY
Glasses for use with the NIGHTSEA BlueStar flashlight meet ANSI Z87.1 impact standards for safety glasses.
Color: Cyan