Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences NIGHTSEA™ Leica EZ4 Fluorescence Adapter DIM Lamp Base, Royal Blue Barrier Filter
SDP

Catalog No. 501929129
Encompass_Preferred

The NIGHTSEA™ Stereo Microscope Fluorescence Adapter adapts the Leica EZ4 stereo microscope for fluorescence with no modification to the microscope itself. The modular design lets you easily switch between six different excitation/emission combinations and white light to work with a variety of fluorescent proteins and other fluorophores. Excellent for quick screening of your fluorescent genotypes, genotype sorting, fluorescence-aided dissection, injection, or micromanipulation, freeing up your research-grade fluorescence microscopes for more demanding work, and bringing fluorescence into the teaching laboratory. Includes lamp base with power, lighthead, ring adapter, barrier filter and filter sheild.
Color: Royal Blue
Excitation: 440-460nm
Emission filter: 500 nm longpass
Fluorophores: GFP, eGFP, fluorescein

Catalog No. 50-192-9129 Supplier Electron Microscopy Sciences Supplier No. SFAZRBDIM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The NIGHTSEA™ Stereo Microscope Fluorescence Adapter adapts the Leica EZ4 stereo microscope for fluorescence with no modification to the microscope itself. The modular design lets you easily switch between six different excitation/emission combinations and white light to work with a variety of fluorescent proteins and other fluorophores. Excellent for quick screening of your fluorescent genotypes, genotype sorting, fluorescence-aided dissection, injection, or micromanipulation, freeing up your research-grade fluorescence microscopes for more demanding work, and bringing fluorescence into the teaching laboratory. Includes lamp base with power, lighthead, ring adapter, barrier filter and filter sheild.
Color: Royal Blue
Excitation: 440-460nm
Emission filter: 500 nm longpass
Fluorophores: GFP, eGFP, fluorescein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.